Learn More
Gosselin™ Conical Containers
Product Code. 11819002
Change view
Click to view available options
Capacity (Metric):
200 mL
400 mL
1000 mL
Color:
Natural
White
Diameter (Metric):
62 mm
85 mm
105 mm
Closure Included:
No
Yes
Yes (Unassembled)
Unit Size:
Pack of 120
Pack of 220
Pack of 250
Pack of 460
Pack of 660
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Diameter (Metric) Closure Included unitSize
11819002 200 mL Natural 62 mm No Pack of 660
11783164 400 mL Natural 85 mm Yes (Unassembled) Pack of 460
11871091 200 mL Natural 62 mm Yes (Unassembled) Pack of 660
12612225 1000 mL Natural 105 mm Yes Pack of 120
11783154 1000 mL Natural 105 mm Yes (Unassembled) Pack of 250
11730084 200 mL Natural 62 mm Yes Pack of 220
11793154 1000 mL Natural 105 mm No Pack of 250
11703174 400 mL Natural 85 mm No Pack of 460
12612205 1000 mL White 105 mm Yes (Unassembled) Pack of 120
11809012 200 mL White 62 mm Yes Pack of 660
11743174 400 mL White 85 mm Yes Pack of 460
11723174 1000 mL White 105 mm Yes Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11819002 Supplier Gosselin™ Supplier No. PC200A05
Add to basket
Add to basket
Provides safe and secure storage in the laboratory. Gosselin™ Conical Containers are manufactured from either clear or white polypropylene.
  • Shock resistant
  • Molded graduations outside on clear containers; molded internal graduations on white containers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Container
Closure Included No
Graduated Yes
Material Polypropylene
Shape Conical, Cylindrical
Sterility Non-sterile
Capacity (Metric) 200 mL
Certifications/Compliance EC N°1935/2004
Color Natural
Diameter (Metric) 62 mm
Graduations 25 mL
Height (Metric) 88 mm
No. per Case 660 /Cs.