Learn More
Gosselin™ Transfer Pipets, PE
Product Code. 12367447
Change view
Click to view available options
Quantity:
5000/Cs.
Unit Size:
Pack of 5000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12367447 5000/Cs. Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12367447 Supplier Gosselin™ Supplier No. PAS300
  • ONLINE EXCLUSIVE PRICE
    207.00 EUR
    132.41 EUR / Pack of 5000
    Save 74.59 EUR
    36% Off
Estimated Shipment: 15-12-2026
Ideal for small-volume liquid transfers. Gosselin™ Transfer Pipets, PE are constructed of polyethylene and graduated for convenient measuring.
  • Polyethylene construction
  • Graduated
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
For Use With (Application) Used in dairy, environment, industries, microbiology
Quantity 5000/Cs.
Packaging Type 500/Box
Color Natural
Volume (Metric) 3 mL
Length (Metric) 150 mm