Learn More
Gosselin™ Square HDPE 150mL Bottles
Description
- Large mouth
- Break resistant
- Sterile or aseptic by irradiation
Specifications
Specifications
| Capacity (Metric) | 150 mL |
| Color | Natural |
| Description | Square |
| Diameter (Metric) Inner Neck | 37 mm |
| Material | HDPE |
| Length (Metric) | 50mm |
| Sterility | Non-sterile |
| Width (Metric) | 50mm |
| Closure Color | Blue |
| Certifications/Compliance | EC N°1935/2004, ISO 11137 |
| Show More |