Learn More
Gosselin™ Pocket ZIP Bags
Product Code. 11723254
Change view
Click to view available options
Color:
Blue
Natural
Yellow
Length (Metric):
170 mm
220 mm
300 mm
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Unit Size
11723254 Natural 170 mm Pack of 1000
11713254 Blue 170 mm Pack of 1000
11743254 Natural 220 mm Pack of 1000
11793254 Natural 300 mm Pack of 1000
11887313 Yellow 170 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 11723254 Supplier Gosselin™ Supplier No. SP218
  • ONLINE EXCLUSIVE PRICE
    113.59 EUR
    70.52 EUR / Pack of 1000
    Save 43.07 EUR
    37% Off
Estimated Shipment: 08-12-2026
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Provides an excellent transport and storage option for solid samples. Gosselin™ Pocket ZIP Bags are nonsterile and manufactured from PE plastic.
  • Quick closure system
  • Single or double pocket
  • Available in a range of colors
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
For Use With (Application) Used in collection and transportation of solid samples and tubes
Material Polyethylene
Quantity 4000 Carton
Length (Metric) 170 mm
Width (Metric) 125 mm
Thickness (Metric) 50 μm