Learn More
Gosselin™ Round-Base Test Tubes
Description
- For laboratory use
- All tubes are CE + IVD marked
- Available with or without closure
- Compatible with automation systems
- Available in sterile or non-sterile (sterility provided by electron beam treatment at 10kGy)
Specifications
Specifications
| Certifications/Compliance | CE, IVD, EC N°1935/2004 |
| Color | Natural |
| Capacity (Metric) | 3 mL |
| Material | Polystyrene |
| For Use With | Clinical, Dairy, Environment, Industries, Microbiology |
| Packaging | 760BG |
| Quantity | 3800 Carton |
| Rimmed | No |
| Sterility | Non-sterile |
| Diameter (Metric) | 12.8 mm |
| Show More |