Learn More
3M™ GoggleGear™ Protective Goggle 3000 Series, PVC Shroud, Clear AF Lens, Premium Cloth Strap
Product Code. 31875401 Shop All 3M Products
Change view
Click to view available options
Frame Material:
PVC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Material Unit Size
31875401 PVC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31875401 Supplier 3M™ Supplier No. 7100325039
  • ONLINE EXCLUSIVE PRICE
    20.65 EUR
    13.08 EUR / Pack of 1
    Save 7.57 EUR
    36% Off
Estimated Shipment: 27-11-2026
Protective goggle 3000 series features a full-size design with wraparound lens for improved field of vision, and slotted nose bridge for a more comfortable fit
  • 3M™ GoggleGear™ Protective Goggle 3000 Series features a full-size design with wrap-around lens for improved field of vision, and slotted nose bridge for a more comfortable fit.
  • Choose from models with anti-fog or 3M™ Scotchgard™ Anti-Fog coating.
  • The GoggleGear™ 3000 is available with indirect venting for enhanced air circulation and anti-fog performance, and is 3, protection against droplets, 4, protection against large particles, and 6, protection against streams of liquid, rated.
  • Includes either an adjustable polyester cloth strap, or easy to clean adjustable neoprene strap, as well as an optimized shroud design with slots to accommodate many prescription eyewear.
  • Acetate lens model meets the chemical resistance requirement of ISO 16321, helping protect workers from chemical hazards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Color Clear
Lens Tint Clear
Product Type Anti-fog Goggles
Disposable Reusable
Lens Material Acetate
Frame Material PVC
UV Protection None
Frame Type Full Frame