Learn More
Goat anti-Canine IgM Fc Secondary Antibody, FITC, Novus Biologicals™
Product Code. 18633358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18633358 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18633358 Supplier Novus Biologicals Supplier No. NBP269669
Goat Polyclonal Antibody
IgM Fc Polyclonal antibody specifically detects IgM Fc in Canine samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgM Fc
Applications ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate FITC
Dilution ELISA, Immunohistochemistry-Frozen, Immunocytochemistry
Formulation Lyophilized from PBS (pH 7.2)
Host Species Goat
Immunogen Purified normal IgM isolated from pooled dog serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Canine
Content And Storage Store at 4°C in the dark.
Form Lyophilized