missing translation for 'onlineSavingsMsg'
Learn More

GNE-493, MedChemExpress

Product Code. 16454178
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454178 10 mg 10mg
16416887 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16454178 Supplier Medchem Express Supplier No. HY10811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNE-493 is a potent, selective, and orally available dual pan-PI3-kinase/mTOR inhibitor with IC50s of 3.4 nM, 12 nM, 16 nM, 16 nM and 32 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR.

GNE-493 is a potent, selective, and orally available dual pan-PI3-kinase/mTOR inhibitor with IC50s of 3.4 nM, 12 nM, 16 nM, 16 nM and 32 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR.

Specifications

Chemical Name or Material GNE-493
Physical Form Solid
CAS 1033735-94-2
Color Dust Gray
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H20N6O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 45 mg/mL (120.82 mM)
SMILES OC(C)(C)C1=CC2=C(S1)C(N3CCOCC3)=NC(C4=CN=C(N)N=C4)=N2
Molecular Weight (g/mol) 372.44
Formula Weight 372.44
Percent Purity 98.33%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.