missing translation for 'onlineSavingsMsg'
Learn More

GNE-140 racemate, MedChemExpress

Product Code. p-200095818
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480689 1 mg 1mg
16473148 10 mg 10mg
16425887 5 mg 5mg
16409706 50 mg 50mg
16404446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480689 Supplier Medchem Express Supplier No. HY100742

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GNE-140 racemate is a racemate mixture of (R)-GNE-140 and (S)-GNE-140. GNE-140 racemate is a potent lactate dehydrogenase A (LDHA) inhibitor.

GNE-140 racemate is a racemate mixture of (R)-GNE-140 and (S)-GNE-140. GNE-140 racemate is a potent lactate dehydrogenase A (LDHA) inhibitor.

Specifications

Chemical Name or Material GNE-140 racemate
CAS 1802977-61-2
Color White
Purity Grade Notes Research
Molecular Formula C25H23ClN2O3S2
Quantity 1 mg
Solubility Information DMSO : 20 mg/mL (40.08 mM; Need ultrasonic)
SMILES O=C(C1SC2=CC=CC=C2Cl)NC(C3=CSC=C3)(C4=CC=C(N5CCOCC5)C=C4)CC1=O
Molecular Weight (g/mol) 499.04
Formula Weight 499.04
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.