missing translation for 'onlineSavingsMsg'
Learn More

Glyphosate-13C2,15N, TRC

Product Code. 17993545
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993545 1 mg 1mg
17913055 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17993545 Supplier Toronto Research Chemicals Supplier No. TRCG7650021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185107-63-4
Molecular Weight (g/mol) 172.05
IUPAC Name 2-(phosphonomethylamino)acetic acid

Specifications

Molecular Formula 13C2 C H8 15N O5 P
Quantity 1 mg
Synonym Glycine-1,2-13C2-15N, N-(phosphonomethyl)- (ACI), N-(Phosphonomethyl)glycine-1,2-13C2-15N (ACI), Glyphosate-13C2,15N, Glyphosate 1,2-13C2 15N, Glyphosate-13C2,15N (glycine-13C2,15N)
InChI Formula InChI=1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1+1,3+1,4+1
SMILES O[13C](=O)[13CH2][15NH]CP(=O)(O)O
Formula Weight 172.0178
Percent Purity >95%
Chemical Name or Material Glyphosate 1,2-13C2 15N
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.