Learn More
Glymidine, TRC
Product Code. 30402447
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402447 2.5 mg
30396638 100 mg
30397261 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30402447 Supplier Toronto Research Chemicals Supplier No. TRCG7520002.5MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 339-44-6
Molecular Formula C13 H15 N3 O4 S
Molecular Weight (g/mol) 309.34
Synonym Benzenesulfonamide, N-[5-(2-methoxyethoxy)-2-pyrimidinyl]-, 2-Benzenesulfonamido-5-(2-methoxyethoxy)pyrimidine, Glidiazine, Glymidine, N-[5-(2-Methoxyethoxy)-2-pyrimidinyl]benzenesulfonamide
IUPAC Name N-[5-(2-methoxyethoxy)pyrimidin-2-yl]benzenesulfonamide
SMILES COCCOc1cnc(NS(=O)(=O)c2ccccc2)nc1
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C13H15N3O4S/c1-19-7-8-20-11-9-14-13(15-10-11)16-21(17,18)12-5-3-2-4-6-12/h2-6,9-10H,7-8H2,1H3,(H,14,15,16)
Formula Weight 309.0783
Chemical Name or Material Glymidine