Learn More
Glycolic Acid, TRC
Product Code. 30134325
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134325 100 g 100g
30136249 10 g 10g
17942935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134325 Supplier Toronto Research Chemicals Supplier No. TRCG645600100G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-14-1
Synonym Acetic acid, 2-hydroxy- (ACI), 2-Hydroxyacetic acid (ACI), Acetic acid, hydroxy- (9CI), Glycolic acid (7CI, 8CI), 2-Hydroxyethanoic acid, Glycocide, GlyPure, GlyPure 70, GlyPure 99, Hydroxyacetic acid, Hydroxyethanoic acid, NSC 166, YST-GCS, α-Hydroxyacetic acid
SMILES OCC(=O)O
Specifications
Molecular Formula C2 H4 O3
InChI Formula InChI=1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)
IUPAC Name 2-hydroxyacetic acid
Molecular Weight (g/mol) 76.05
Formula Weight 76.016
Percent Purity >95%
Quantity 100 g
Chemical Name or Material Glycolic acid
Grade GC