Learn More
Glycolic Acid, Supelco™
Specifications
Specifications
| CAS | 79-14-1 |
| Melting Point | 74°C to 78°C |
| Boiling Point | 100°C (Decomposition) |
| Density | 1.49 g/cm3 at 25°C |
| Quantity | 100 g |
| MDL Number | MFCD00004312 |
| Synonym | Glycolic acid, Hydroxyacetic acid, Glycolic acid |
| Molecular Weight (g/mol) | 76.05 |
| Chemical Name or Material | Glycolic Acid |