Learn More
Glycolic Acid, Sigma-Aldrich™
Product Code. 15824301
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15824301 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15824301 Supplier Sigma Aldrich Supplier No. 8141861000
 Restricted Item
Estimated Shipment: 16-11-2026
70% Solution in water for synthesis
TRUSTED_SUSTAINABILITY
Specifications
pH 0.1 (25°C in H2O)
Melting Point 10°C
Boiling Point 112°C/1013 hPa
Density 1.262 g/cm3 at 20°C
Quantity 1 L
Chemical Name or Material Glycolic Acid