missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycolic acid oxidase inhibitor 1, MedChemExpress
Description
Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1, in Table IV.
Specifications
Specifications
| Chemical Name or Material | Glycolic acid oxidase inhibitor 1 |
| CAS | 77529-42-1 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C16H10BrNO3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 137 mg/mL (398.07 mM) |
| SMILES | O=C(C(C1=CC=C(C2=CC=C(Br)C=C2)C=C1)=C3O)NC3=O |
| Molecular Weight (g/mol) | 344.16 |
| Formula Weight | 344.16 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?