Learn More
Hach Lange™ Glycol Test Reagent 1 Powder Pillows
Brand:  Hach Lange™ 2183668
Product Code. 10386501
  • 18.74 EUR / Pack of 1
 Restricted Item
Estimated Shipment: 02-12-2026
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Ethylene Glycol 1 Reagent
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to Disposal.
Supplemental information
  • MIXTURE LIST-Contain: Potassium (meta)periodate