Learn More
Glycochenodeoxycholic Acid Sodium, 95%, TRC
Specifications
Specifications
| Chemical Name or Material | Glycochenodeoxycholic Acid Sodium Salt |
| CAS | 16564-43-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C26 H42 N O5 . Na |
| Quantity | 25 mg |
| Synonym | N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Sodium, N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Monosodium, NSC 681056, Sodium Chenodeoxycholylglycine, Sodium Glycochenodeoxycholate, Sodium Glycochenodesoxycholate |
| InChI Formula | InChI=1S/C26H43NO5.Na/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29;/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21-,24+,25+,26-;/m1./s1 |
| SMILES | [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate |
| Molecular Weight (g/mol) | 471.61 |
| Show More |