missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monoricinoleate, MP Biomedicals

Product Code. 11453245 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453245 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453245 Supplier MP Biomedicals Supplier No. 0521401680

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1323-38-2
Molecular Formula C21H40O5
Quantity 100 g
Synonym glyceryl monoricinoleate, glyceryl ricinoleate, glycerol ricinoleate, 9-octadecenoic acid, 12-hydroxy-, r-z-, monoester with 1,2,3-propanetriol, ricinoleic acid 2,3-dihydroxypropyl ester, 2,3-dihydroxypropyl 9z-12-hydroxy-9-octadecenoate, 12-hydroxy-,monoesterwith1,2,3-propanetriol, r-z-9-octadecenoicacid, 9-octadecenoicacid,12-hydroxy-, r-z-,monoesterwith1,2,3-propanetriol
InChI Key HDIFHQMREAYYJW-ZJNNSCJFSA-N
SMILES CCCCCCC(CC=CCCCCCCCC(=O)OCC(CO)O)O
IUPAC Name 2,3-dihydroxypropyl (E,12R)-12-hydroxyoctadec-9-enoate
Molecular Weight (g/mol) 372.546
PubChem CID 101321425
Chemical Name or Material Glyceryl Monoricinoleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.