Learn More
Glycerol Triacetate, Millipore™
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product can replace highly toxic diluents in membrane preparation and thus aligns with \Less Hazardous Chemical Syntheses\.
Specifications
Specifications
| CAS | 102-76-1 |
| Melting Point | -78°C |
| Density | 1.16 g/cm3 at 20°C |
| Boiling Point | 258°C/1013 hPa |
| MDL Number | MFCD00008716 |
| Quantity | 500 mL |
| Synonym | Glycerol triacetate, Glycerol triacetate |
| Molecular Weight (g/mol) | 218.2 |
| Grade | Research |