missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, Sigma-Aldrich™

Product Code. 15860571
Change view
Click to view available options
Quantity:
1 L
5 L
25 L
Unit Size:
1 L
25 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860571 25 L 25 L
15846590 1 L 1 L
15856590 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15860571 Supplier Sigma Aldrich Supplier No. 8187099025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anhydrous for synthesis

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. Glycerol, with its high boiling point, low flammability, and polarity, is a standout choice. As a non-toxic and biodegradable compound, it aligns perfectly with the principles of ′Safer Solvents and Auxiliaries′ and ′Design for Degradation′.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5

Specifications

Melting Point 20°C (lit.)
Density 1.25 g/mL
Quantity 25 L
Molecular Formula HOCH2CH(OH)CH2OH
MDL Number MFCD00004722
Synonym 1,2,3-Propanetriol, Trihydroxylpropane, Protol, Glycerin, 1,2,3-Propanetriol, Glycerin, 1,2,3-Propanetriol, Glycerin
Boiling Point 182°C/20 mmHg
Molecular Weight (g/mol) 92.09
Grade Research
Chemical Name or Material Glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.