missing translation for 'onlineSavingsMsg'
Learn More

Glycerol-3-phosphate oxidase, MP Biomedicals™

Product Code. 11499661 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 U
50 U
500 U
Unit Size:
100 units
50 units
500 units
This item is not returnable. View return policy

Product Code. 11499661

Brand: MP Biomedicals 0215724080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9046-28-0
Synonym sn-Glycerol-3-phosphate, oxygen 2-oxidoreductase

Specifications

Color Yellow
Quantity 100 U
Physical Form Lyophilized Powder
Chemical Name or Material Glycerol-3-phosphate oxidase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.