missing translation for 'onlineSavingsMsg'
Learn More

Glycerol-13C3, TRC

Product Code. 30137297
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137297 10 mg 10mg
30136259 500 mg 500mg
30011806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137297 Supplier Toronto Research Chemicals Supplier No. TRCG59840310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63346-81-6
Molecular Formula 13C3 H8 O3
Molecular Weight (g/mol) 95.07
Synonym 1,2,3-Propanetriol-1,2,3-13C3 (9CI, ACI), 1,2,3-Propanetriol-13C3, Glycerol-13C3
IUPAC Name (1,2,3-^{13}C_{3})propane-1,2,3-triol
SMILES O[13CH2][13CH](O)[13CH2]O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i1+1,2+1,3+1
Formula Weight 95.0574
Percent Purity >95%
Chemical Name or Material Glycerol-13C3
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.