missing translation for 'onlineSavingsMsg'
Learn More

Glutardialdehyde, Sigma-Aldrich™

Product Code. 15849910
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849910 1 L 1 L
15839910 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15849910 Supplier Sigma Aldrich Supplier No. 8143931000
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

50% Solution in water for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-30-8

Specifications

Boiling Point 100.7°C/1013 hPa
Melting Point -33°C
Molecular Formula OHC(CH2)3CHO
MDL Number MFCD00007025
Density 1.13 g/cm3 at 20°C
Synonym Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial, Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial
Molecular Weight (g/mol) 100.12
Grade Reagent
Quantity 1 L
Chemical Name or Material Glutardialdehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.