missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, MP Biomedicals™

Product Code. 11432731 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432731 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11432731 Supplier MP Biomedicals Supplier No. 0219859510
  • ONLINE EXCLUSIVE PRICE
    57.35 EUR
    54.60 EUR / 10mL
    Save 2.75 EUR
    4% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 111-30-8
Color Colorless
Density 1.061g/cm3 at 20°C
Molecular Formula C5H8O2
MDL Number MFCD00007025
Quantity 10 mL
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES O=CCCCC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.12
PubChem CID 3485
ChEBI CHEBI:64276
Formula Weight 100.11
Physical Form Liquid
Chemical Name or Material Glutaraldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.