missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, 25% aqueous solution, MP Biomedicals™

Product Code. 11451102 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451102 10 mL 10mL
11461102 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451102 Supplier MP Biomedicals Supplier No. 0219519910
  • ONLINE EXCLUSIVE PRICE
    190.00 EUR
    180.60 EUR / 10mL
    Save 9.40 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Color Colorless
Density 1.061g/mL
Quantity 10 mL
Formula Weight 100.11
Physical Form Liquid
Chemical Name or Material Glutaraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.