missing translation for 'onlineSavingsMsg'
Learn More

ʟ-Glutamine, Millipore™

Product Code. 32237624
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32237624 10 L 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32237624 Supplier Millipore Supplier No. 1002899010
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For biochemistry

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ʟ-Glutamine
Melting Point 185°C (dec.) (lit.)
CAS 56-85-9
Molecular Formula H2NCOCH2CH2CH(NH2)CO2H
Synonym L-Glutamic acid-5-amide, Gln, (S)-2,5-Diamino-5-oxopentanoic acid, ʟ-Glutamic acid 5-amide, Levoglutamide, (S)-2,5-Diamino-5-oxopentanoic acid, ʟ-Glutamic acid 5-amide, Levoglutamide
Density 1.47 g/cm³ at 20°C
Molecular Weight (g/mol) 146.14
Quantity 10 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.