missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Glutamic Pyruvic Transaminase from Porcine Heart
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
500 U
Unit Size:
1ku
500 units
Description
A transaminase enzyme, glutamic pyruvic transaminase catalyzes the transfer of an amino group from alanine to α-ketoglutarate, the products of this reversible transamination reaction being pyruvate and glutamate.
- Presentation: Off-white powder
- Foreign Activity: GLDH, GOT, LDH, MDH: ≤0.01%
- Specific Activity: ∽80u/mg protein
- Unit Definition: One unit will cause the transamination of 1.0μmole of L-alanine per minute at pH 7.5 and 25°C
- Solubility: At 5mg/mL in 0.05M potassium phosphate pH 7.5, giving a clear, pale-green solution; also soluble in distilled water or dilute buffer
Specifications
Specifications
| Concentration | ≈80 U/mg |
| Color | White |
| pH | 7.5 |
| For Use With (Application) | Used as a marker of liver disease due to the release of GPT into the Blood following liver cell necrosis |
| Content And Storage | -20°C |
| Source | Procine heart |
| Quantity | 1 kU |
| Product Type | Glutamic Pyruvic Transaminase |
| Molecular Weight (g/mol) | 115000 |
| Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction