missing translation for 'onlineSavingsMsg'
Learn More

Glufosinate Ammonium, TRC

Product Code. 30393925
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393925 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393925 Supplier Toronto Research Chemicals Supplier No. TRCG596950250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glufosinate ammonium
CAS 77182-82-2
Purity Grade Notes HPLC
Molecular Formula C5 H11 N O4 P . H4 N
Quantity 250 mg
Synonym Butanoic acid, 2-amino-4-(hydroxymethylphosphinyl)-, monoammonium salt (9CI), Ammonium glufosinate, BASTA 15, Basta, Basta 150SL, Basta Fl, Basta LS, Buster, Dash, Finale, Finale 14SL, Glufosinate ammonium salt, Glufosinate monoammonium salt, Glufosinate-ammonium, HOE 00661, HOE 39866, Ignite, Ignite 280, Liberty, Liberty (pesticide), Liberty 280SL
InChI Formula InChI=1S/C5H12NO4P.H3N/c1-11(9,10)3-2-4(6)5(7)8;/h4H,2-3,6H2,1H3,(H,7,8)(H,9,10);1H3
SMILES [NH4+].CP(=O)([O-])CCC(N)C(=O)O
IUPAC Name azanium;(3-amino-3-carboxypropyl)-methylphosphinate
Molecular Weight (g/mol) 198.16
Formula Weight 198.0769
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.