missing translation for 'onlineSavingsMsg'
Learn More

Glucosylceramidase/GBA Antibody (4H4), Novus Biologicals™

Rabbit Monoclonal Antibody

343.00 EUR - 533.00 EUR

Specifications

Antigen Glucosylceramidase/GBA
Clone 4H4
Dilution Western Blot 1:500-1:5000, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043397
View Documents
Novus Biologicals
NBP3-26559-50ul
50 μL
343.00 EUR
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043788
View Documents
Novus Biologicals
NBP3-26559-100ul
100 μL
533.00 EUR
100µL
Se alternative produkter
Spar op til 
»
Please to purchase this item. Need a web account? Register with us today!
Beskrivelse

Beskrivelse

Glucosylceramidase/GBA Monoclonal antibody specifically detects Glucosylceramidase/GBA in Human samples. It is validated for Western Blot, ELISA
Tekniske data

Tekniske data

Glucosylceramidase/GBA
Western Blot 1:500-1:5000, ELISA
Monoclonal
Purified
RUO
Human
Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta, acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase
A synthesized peptide derived from Human Glucosylceramidase/GBA [UniProt P04062]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
4H4
Western Blot, ELISA
Unconjugated
Rabbit
Lipid and Metabolism
PBS, pH 7.4, 150mM NaCl and 50% glycerol
2629
IgG
Affinity purified
Videoer
SDS
Dokumenter

Dokumenter

Produktcertifikater
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.