Learn More
Glucosylceramidase/GBA Antibody (4H4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26559-100ul
This item is not returnable.
View return policy
Description
Glucosylceramidase/GBA Monoclonal antibody specifically detects Glucosylceramidase/GBA in Human samples. It is validated for Western Blot, ELISA
Specifications
| Glucosylceramidase/GBA | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4H4 | |
| Western Blot 1:500-1:5000, ELISA | |
| Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta, acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase | |
| A synthesized peptide derived from Human Glucosylceramidase/GBA [UniProt P04062] | |
| 100 μL | |
| Lipid and Metabolism | |
| 2629 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |