missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glucose Oxidase Monoclonal Antibody (GO14-6.6.1.2), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

424.00 EUR - 1052.00 EUR

Specifications

Antigen Glucose Oxidase
Clone GO14-6.6.1.2
Concentration 1.5 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17880456
View Documents
Invitrogen™
MA5-46022
100 μg
424.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11558892
GREENER_CHOICE
View Documents
Invitrogen™
MA110605
500 μg
560.00 EUR
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17800256
View Documents
Invitrogen™
MA5-46001
1 mg
1052.00 EUR
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MA1-10605 detects Glucose Oxidase from Aspergillus niger samples. MA1-10605 has been successfully used in ELISA applications. The MA1-10605 immunogen is purified glucose oxidase from Aspergillus niger.

Glucose Oxidase Monoclonal antibody specifically detects Glucose Oxidase in Fungi samples. It is validated for ELISA.
Specifications

Specifications

Glucose Oxidase
1.5 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
Purified glucose oxidase from Aspergillus niger.
Primary
-20°C, Avoid Freeze/Thaw Cycles
GO14-6.6.1.2
ELISA
Unconjugated
Mouse
Fungi
aspergillus niger glucose oxidase; GOx
IgG1
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.