missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glucose Oxidase Monoclonal Antibody (GO14-6.6.1.2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-46001
This item is not returnable.
View return policy
Description
MA1-10605 detects Glucose Oxidase from Aspergillus niger samples. MA1-10605 has been successfully used in ELISA applications. The MA1-10605 immunogen is purified glucose oxidase from Aspergillus niger.
Glucose Oxidase Monoclonal antibody specifically detects Glucose Oxidase in Fungi samples. It is validated for ELISA.
Specifications
| Glucose Oxidase | |
| Monoclonal | |
| 1.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Fungi | |
| Antibody | |
| IgG1 |
| ELISA | |
| GO14-6.6.1.2 | |
| Unconjugated | |
| aspergillus niger glucose oxidase; GOx | |
| Purified glucose oxidase from Aspergillus niger. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction