missing translation for 'onlineSavingsMsg'
Learn More

Gluconic Acid Sodium Salt, Sigma-Aldrich™

Product Code. 15871641
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
50 kg
Unit Size:
100g
1kg
2.5kg
50kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871641 50 kg 50kg
15841641 100 g 100g
15851641 1 kg 1kg
15861641 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15871641 Supplier Sigma Aldrich Supplier No. 8220589050
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-07-1

Specifications

Melting Point 170°C to 175°C
Quantity 50 kg
MDL Number MFCD00064210
Synonym Gluconic acid sodium salt, Sodium gluconate, Gluconic acid sodium salt
Molecular Weight (g/mol) 218.14
Grade Ungraded
Chemical Name or Material Gluconic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.