missing translation for 'onlineSavingsMsg'
Learn More

Gluconic Acid Potassium Salt, Sigma-Aldrich™

Product Code. 15831061
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15831061 500 g 500g
15821061 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15831061 Supplier Sigma Aldrich Supplier No. 8206010500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

Gluconic acid potassium salt commonly known as potassium gluconate is used as chelating agent in organic synthesis. Additionally, it is also used as buffering agent in biochemical and pharmaceutical formulations, to maintain pH stability during reactions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-27-4

Specifications

Melting Point 174°C to 176°C
Quantity 500 g
MDL Number MFCD00064211
Synonym Gluconic acid potassium salt, Potassium gluconate, Gluconic acid potassium salt
Molecular Weight (g/mol) 234.25
Grade Ungraded
Chemical Name or Material Gluconic Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.