Learn More
MedChemExpress [Glu1]-Fibrinopeptide B
Product Code. 16452769
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452769 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16452769 Supplier MedChemExpress Supplier No. HYP0308
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress [Glu1]-Fibrinopeptide B is derived from fibrinopeptide B amino acid residues 1-14. Human fibrinopeptide B (hFpB), a thrombin-derived proteolytic cleavage product of the fibrinogen B beta-chain, to stimulate neutrophils (PMN), monocytes, and fibroblasts.
Specifications
CAS 103213-49-6
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1570.6 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature