Learn More
Globe Scientific Coagulation Cup for Accu Coa Data Coagulation Analyzers
Shop All Globe Scientific ProductsDescription
- Made of Polystyrene
- Supplied with metal mixing bars
- Dimensions: 1.3 x 0.3 in. (35.1 x 8.6mm)
Specifications
Specifications
| Type | Coagulation Cup |
| For Use With (Equipment) | Accu Coa Data Coagulation Analyzers |
| Includes | Metal mixing bars |