Learn More
Globe Scientific Polypropylene Sample Cup for Alpha Wassermann™ ACE and Gilford™ Analyzers
Product Code. 31922712 Shop All Globe Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Alpha Wassermann ACE and Gilford Analyzers
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31922712 Alpha Wassermann ACE and Gilford Analyzers Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31922712 Supplier Globe Scientific Supplier No. 5545
Estimated Shipment: 04-12-2026
For use with Alpha Wassermann ACE and Gilford analyzers.
  • Capacity: 0.5mL
  • Dimensions: 0.3 x 0.8 in. (10 x 22mm)
Specifications
Type Sample Cup
For Use With (Equipment) Alpha Wassermann ACE and Gilford Analyzers
Capacity (Metric) 5 mL
Quantity 1000/Pk.