Learn More
Glipizide, Mikromol™
Description
Glipizide
Specifications
Specifications
| Chemical Name or Material | Glipizide |
| CAS | 29094-61-9 |
| Analyte or Component Names | Glipizide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N5 O4 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | N-[2-[4[[[(Cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methylpyrazinecarboxamide, 1-Cyclohexyl-3-[[p-[2-(5-methylpyrazinecarboxamido) |
| InChI Formula | InChI=1S/C21H27N5O4S/c1-15-13-24-19(14-23-15)20(27)22-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,22,27)(H2,25,26,28) |
| SMILES | Cc1cnc(cn1)C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3 |
| IUPAC Name | N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide |
| Show More |