Learn More
Heidolph™ Glassware Set - Vertical Condenser with Valve (Reflux Distillation)
Product Code. 10657812 Shop All Heidolph Instruments Products
Change view
Click to view available options
Material (Coating):
Transparent plastic
Uncoated
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Coating) Unit Size
10657812 Transparent plastic Each
10055741 Uncoated Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10657812 Supplier Heidolph™ Supplier No. 5130061000
Estimated Shipment: 28-12-2026
Vertical condenser, centerpiece with valve for reflux distillation
  • Glassware sets with a transparent plastic coating are suitable for a temperature range up to max. 80™C
  • Glassware set includes (1) 1000 mL evaporating flask and (1) 1000 mL receiving flask
  • Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29
  • Ventilation cap without ground joints for easy handling and grease-free operation included
  • A venting valve with refill function is optionally available
  • Not suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
  • Also available upon request: glassware sets for Soxhlet extraction and descending condenser system
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Glassware Set
For Use With (Equipment) Rotary Evaporators
Includes Glassware set G6B - Vertical condenser centerpiece with valve (reflux distillation), 1000 mL evaporating flask and 1000 mL receiving flask with transparent plastic coating
Material (Coating) Transparent plastic
Volume (Metric) 1000 mL