Learn More
Poulten Selfe™ Glass Ubbelohde
Description
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.
Specifications
Specifications
| Product Type | Ubbelohde Viscometer |
| Viscosity Range | 100 to 500 cSt |
| Calibrated | Yes |
| Material | Glass |
| Size | 2B |
Product Title
Haben Sie Verbesserungsvorschläge?