Learn More
Poulten Selfe™ Glass U-Tube Reverse Flow Viscometer
Product Code. 13121043
Change view
Click to view available options
Size:
9
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
13121043 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13121043 Supplier Poulten Selfe™ Supplier No. 1637/09
  • ONLINE EXCLUSIVE PRICE
    236.00 EUR
    224.09 EUR / Pack of 1
    Save 11.91 EUR
    5% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Suitable viscometer for measuring transparent and opaque liquids. Poulten Selfe™ Glass U-Tube Reverse Flow Viscometer
  • Type BS/IP/RF
  • BS 188, ASTM D445-IP71, ASTM D2170, ISO 3104, ISO
  • Requires long filling pipette
Specifications
Product Type U-Tube Reverse Flow Viscometers
Viscosity Range 6000 to 30,000 cSt
Calibrated Yes
Description Kinematic viscosity: 6,000 to 30,000mm2/s (cSt)
Material Glass
Size 9
Volume (Metric) 12 - 25 mL