Learn More
Karl Hecht™ Glass Tank
Product Code. 10797302
Change view
Click to view available options
Dimensions (L x W x H):
36 x 23 x 26 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
10797302 36 x 23 x 26 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10797302 Supplier Karl Hecht™ Supplier No. 40070006
  • ONLINE EXCLUSIVE PRICE
    139.65 EUR
    98.09 EUR / Pack of 1
    Save 41.56 EUR
    29% Off
Estimated Shipment: 23-11-2026
Suitable for general laboratory applications.
  • Fine ground rim
  • Made of clear glass for excellent optical clarity
  • 23 x 26 x 36cm
Specifications
Material Glass
For Use With (Application) General laboratory applications
Includes Fine ground rim
Dimensions (L x W x H) 36 x 23 x 26 cm