Learn More
Karl Hecht™ Assistent™ Clear Glass Beads
Product Code. 10469401
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10469401 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10469401 Supplier Karl Hecht™ Supplier No. 41401005
  • ONLINE EXCLUSIVE PRICE
    41.54 EUR
    34.66 EUR / 1kg
    Save 6.88 EUR
    16% Off
Estimated Shipment: 09-12-2026
Excellent for spreading the boiling area or for mixing and packing applications in the laboratory. Karl Hecht™ Assistent™ Clear Glass Beads are polished for exceptional smoothness.
  • Spherical
  • Solid, good hardness
  • Corrosion-free and resistant to thermal shock
Order Info
dimType
Specifications
Type Shot
Material Glass
Quantity 1 kg