Learn More
Hirschmann™ Class AS Schellbach Blue Graduated Micro Burettes
Product Code. 11722392
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11722392 2 mL Pack of 2
11732392 5 mL Pack of 2
11742392 10 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11722392 Supplier Hirschmann™ Supplier No. 3300240
  • ONLINE EXCLUSIVE PRICE
    493.00 EUR
    402.00 EUR / Pack of 2
    Save 91.00 EUR
    18% Off
Estimated Shipment: 05-01-2027
With Schellbach stripes for enhanced legibility.
  • Class AS
  • Manufactured from DURAN™ borosilicate glass for resistance to chemicals and thermal shock
  • Blue graduation marks
  • Certified conformity, with dated batch identification
  • Straight stopcock with PTFE key
  • Stopcock in filling tube with PTFE key
  • Main point ring graduation
  • Calibrated for EX
  • Acc. to DIN EN ISO 385
Specifications
Capacity (Metric) 2 mL
Material Glass
Graduations 0.01 mL
Includes Schellbach Stripes
Plug Material PTFE
Scale Blue
Stopcock Straight
Stopcock Material PTFE
Tolerance ±0.01 mL
Class Class AS
Certifications/Compliance DIN EN ISO 385