Learn More
Merck Glass Funnel for Vacuum Filtration
Product Code. 15885654 Shop All Merck Products
Change view
Click to view available options
Capacity (Metric):
300 mL
1 L
Description:
Ground glass seal
PTFE-faced
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Description unitSize
15885654 300 mL PTFE-faced Each
15869512 300 mL Ground glass seal Each
15884156 1 L Ground glass seal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15885654 Supplier Merck Supplier No. XX1014724
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Millipore Glass Vacuum Filtration Accessory ProductThese accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration. This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits, Millipore 47 mm All-Glass Filter Holder Kits, and Millipore MilliSolve Kit.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 47 mm
Capacity (Metric) 300 mL
Material PTFE-faced Borosilicate Glass
Color Clear
Description PTFE-faced