Learn More
Fisherbrand™ Glass-Encased Stir Bar
Product Code. 11567792 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
6 mm
12 mm
19 mm
25 mm
45 mm
60 mm
Diameter (Metric):
5 mm
6 mm
8 mm
Unit Size:
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
11567792 25 mm 6 mm Pack of 5
11537792 6 mm 5 mm Pack of 5
11848902 45 mm 8 mm Pack of 5
11858902 60 mm 8 mm Pack of 5
11547792 12 mm 5 mm Pack of 5
11557792 19 mm 5 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11567792
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
Use with very abrasive media that may erode PTFE. The glass-encased magnetic stir bar is cylindrical for a wide variety of uses.
  • Stable, general-purpose magnetic stir bar
  • Use with very abrasive media that may erode PTFE
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color Clear
Description Glass encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 6 mm
Length (Metric) 25 mm
Quantity 5/Pk.
Shape Cylinder