Learn More
Brand™ Graduated Glass Centrifuge Tube
Product Code. 12015352 Shop All Brand Products
Change view
Click to view available options
Graduations:
0 to 10 mL
0 to 15 mL
Material:
AR-GLAS™
Borosilicate Glass (3.3)
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Graduations Material unitSize
12015352 0 to 15 mL AR-GLAS™ Pack of 10
12005352 0 to 10 mL AR-GLAS™ Pack of 10
11858261 0 to 10 mL Borosilicate Glass (3.3) Pack of 10
11868261 0 to 15 mL Borosilicate Glass (3.3) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12015352 Supplier Brand™ Supplier No. 778712
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Graduations and inscriptions in high contrast white enamel.
  • Manufactured from either AR-GLAS™ or Boro 3.3
  • Capacity of 15mL
  • Provided in packs of 10
  • Graduation subdivision of 0.1mL on all models
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 15 mL
Material AR-GLAS™
No. per Case 10 Pack
No. per Pack 10
Quantity 10/Pk.
Bottom Shape Conical
Diameter (Metric) 17 mm
Graduations 0 to 15 mL
Product Type Centrifuge Tube
Max. RCF 3500 x g