Learn More
Jaytec Glass™ Glass Capillary Tubes
Product Code. 12352078
Change view
Click to view available options
Length (Metric) Tube:
100 mm
Diameter (Metric) Outer:
1 mm
Volume (Metric):
50 μL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tube Diameter (Metric) Outer Volume (Metric) Unit Size
12352078 100 mm 1 mm 50 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12352078 Supplier Jaytec Glass™ Supplier No. CAP10010
Estimated Shipment: 10-12-2026
These Glass Capillary Tubes use a liquid's properties of cohesion, adhesion and surface tension to draw liquid up the interior walls of a narrow tube.
  • Manufactured from industry-standard soda glass
Specifications
Material Soda Glass
Volume (Metric) 50 μL
Product Type Capillary Tubes
Diameter (Metric) Outer 1 mm
Length (Metric) Tube 100 mm
Quantity 1000 Pk.