Learn More
Hirschmann™ ringcaps™ Disposable Capillary Pipettes
Product Code. 15539035
Change view
Click to view available options
Volume (Metric):
40 μL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15539035 40 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15539035 Supplier Hirschmann™ Supplier No. 9600140
  • ONLINE EXCLUSIVE PRICE
    136.00 EUR
    114.20 EUR / Pack of 1000
    Save 21.80 EUR
    16% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
  • With ringmark
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Accuracy 0.25%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material Glass
Includes Ring Mark
Volume (Metric) 40 μL
Type Capillary Pipette