Learn More
MP Biomedicals™ Glass Beads
Product Code. 11481505 Shop All MP Biomedicals Products
Change view
Click to view available options
Diameter (Metric):
4 mm
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11481505 4 mm 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11481505 Supplier MP Biomedicals™ Supplier No. 112521011
  • ONLINE EXCLUSIVE PRICE
    241.00 EUR
    228.90 EUR / 250g
    Save 12.10 EUR
    5% Off
Estimated Shipment: 18-12-2026